1,4-Dibromoisoquinoline 95%
Supplier: Ambeed
|
Warning
|
A134787-10G
A134787-250MG
A134787-1G
A134787-5G
103802-052EA
42
USD
103802-052
77109-454
77109-456
77314-828
77314-830
1,4-Dibromoisoquinoline 95%
1,4-Dibromoisoquinoline
Precaución: for Research Use Only
Formula:
C₉H₅Br₂N MW: 286.9507 g/mol Boiling Pt: 352.5 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00234475 CAS Number: 51206-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...