1,4-Dibromo-2-iodobenzene 97%

Supplier: Ambeed
Warning

A242170-25G A242170-100G A242170-10G A242170-1G
103811-458EA 20 USD
103811-458 103851-270 77090-440 77193-368 77193-370 77304-242
1,4-Dibromo-2-iodobenzene 97%
1,4-Dibromo-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂I
Boiling Pt: 303.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07778993
CAS Number: 89284-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR