2,5-Dibromo-1-fluorobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dibromo-2-fluorobenzene

A101583-100G A101583-10G A101583-25G A101583-500G
103821-724EA 6 USD
103821-724 77148-220 77148-222 77148-224 77148-226
2,5-Dibromo-1-fluorobenzene 98%
2,5-Dibromo-1-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂F
MW: 253.9 g/mol
Boiling Pt: 216
Melting Pt: 35
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00010603
CAS Number: 1435-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR