1,4-Dibromo-2-fluoro-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A469133-100G A469133-10G A469133-250MG A469133-1G A469133-25G A469133-5G
103828-006EA 7 USD
103828-006 103839-080 76818-496 76818-498 76818-500 76842-986 77307-464 77307-466
1,4-Dibromo-2-fluoro-5-nitrobenzene 97%
1,4-Dibromo-2-fluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD17014707
CAS Number: 366496-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR