2-Chloro-1,4-dibromobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Dibromochlorobenzene, 1,4-Dibromo-2-chlorobenzene, 1-Chloro-2,5-dibromobenzene
A314496-25G
A314496-5G
A314496-100G
A314496-10G
103835-958EA
6
USD
103835-958
76838-774
76838-776
77194-716
77194-718
2-Chloro-1,4-dibromobenzene 98%
2-Chloro-1,4-dibromobenzene
Precaución: for Research Use Only
Formula:
C₆H₃Br₂Cl MW: 270.35 Boiling Pt: 259 Melting Pt: 39…42 °C Flash Pt: 119.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018094 CAS Number: 3460-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...