2,5-Dibromo-p-xylene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Dibromo-1,4-dimethylbenzene, 1,4-Dibromo-2,5-dimethylbenzene
A157397-100G
A157397-500G
A157397-5G
A157397-1G
A157397-25G
103805-002EA
5
USD
103805-002
77247-534
77247-536
77247-538
77319-020
77319-022
2,5-Dibromo-p-xylene 98%
2,5-Dibromo-p-xylene
Precaución: for Research Use Only
Formula:
C₈H₈Br₂ MW: 263.96 g/mol Boiling Pt: 261 Melting Pt: 75 Flash Pt: 124.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000091 CAS Number: 1074-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...