2,5-Dibromo-1,4-dimethoxybenzene 98%

Supplier: Ambeed
Warning

A217981-25G A217981-5G A217981-100G A217981-10G A217981-1G
103818-678EA 6 USD
103818-678 103849-410 77088-060 77088-062 77097-884 77097-886 77303-470
2,5-Dibromo-1,4-dimethoxybenzene 98%
2,5-Dibromo-1,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O₂
Boiling Pt: 290.3 °C (760 mm Hg)
Melting Pt: 144…147 °C
Flash Pt: 116.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00025694
CAS Number: 2674-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR