1,4-Dibromo-2,5-difluorobenzene 98%
Supplier: Ambeed
|
Warning
|
A191638-1000G
A191638-25G
A191638-5G
A191638-100G
A191638-10G
A191638-500G
103809-260EA
337
USD
103809-260
103846-992
77084-840
77084-842
77084-844
77287-486
77287-488
77287-490
1,4-Dibromo-2,5-difluorobenzene 98%
1,4-Dibromo-2,5-difluorobenzene
Precaución: for Research Use Only
Formula:
C₆H₂Br₂F₂ MW: 271.89 g/mol Boiling Pt: 209 Melting Pt: 65 Storage Temperature: Ambient |
MDL Number:
MFCD00000346 CAS Number: 327-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...