1,4-Dibromo-2,3-dimethylbenzene 97%

Supplier: Ambeed
Warning

A293474-10G A293474-250MG A293474-1G A293474-25G A293474-5G
103813-984EA 10 USD
103813-984 103835-122 76816-620 76816-622 77305-630 77305-632 77305-634
1,4-Dibromo-2,3-dimethylbenzene 97%
1,4-Dibromo-2,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Melting Pt: 42…46 °C
Storage Temperature: Ambient
MDL Number: MFCD06796460
CAS Number: 75024-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR