1,4-Dibromo-2,3-difluorobenzene 98%

Supplier: Ambeed
Warning

A105757-100G A105757-10G A105757-25G A105757-1G A105757-5G
103796-932EA 23 USD
103796-932 103822-674 77188-768 77188-770 77188-772 77309-540 77309-542
1,4-Dibromo-2,3-difluorobenzene 98%
1,4-Dibromo-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂F₂
MW: 271.8849 g/mol
Boiling Pt: 212 °C (760 mm Hg)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09835192
CAS Number: 156682-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR