2,5-Dibromo-α,α,α-trifluorotoluene 98%

Supplier: Ambeed
Warning

A303214-5G A303214-10G
103835-530EA 7 USD
103835-530 76838-216 77250-824
2,5-Dibromo-α,α,α-trifluorotoluene 98%
2,5-Dibromo-α,α,α-trifluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃Br₂F₃
Melting Pt: 49…51 °C
Storage Temperature: Ambient
MDL Number: MFCD00013554
CAS Number: 7657-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR