1,4-Diazabicyclo[3.2.2]nonane 97%

Supplier: Ambeed
Warning

Synonyms: N-phenyl-1,4-diazabicyclo[3.2.2]nonane-4-carboxamide, 1,4-diaza-bicyclo[3.2.2]nonane, 1,4-Diazobicylco[3.2.2]nonane

A198807-10G A198807-250MG A198807-1G A198807-5G
103817-048EA 91 USD
103817-048 103847-676 77030-076 77030-078 77085-698 77302-602
1,4-Diazabicyclo[3.2.2]nonane 97%
1,4-Diazabicyclo[3.2.2]nonane

Precaución: for Research Use Only

Formula: C₇H₁₄N₂
MW: 126.2 g/mol
Boiling Pt: 180 °C (760 mm Hg)
Flash Pt: 71.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14581694
CAS Number: 283-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR