1,4-Cyclohexanedione 98%

Supplier: Ambeed
Warning

A105913-100G A105913-10G A105913-1G A105913-25G A105913-500G A105913-5G
103807-944EA 127 USD
103807-944 103822-716 77107-624 77107-626 77107-628 77107-630 77107-632 77283-412
1,4-Cyclohexanedione 98%
1,4-Cyclohexanedione

Precaución: for Research Use Only

Formula: C₆H₈O₂
Boiling Pt: 226.6 °C (760 mm Hg)
Melting Pt: 77…78.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00001606
CAS Number: 637-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR