[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride 97%

Supplier: Ambeed
Warning

Synonyms: 1,4-Butylenebis(diphenylphosphine)palladium dichloride, Dichloro[1,4-bis(diphenylphosphino)butane]palladium(II)

A327496-1G A327496-5G A327496-250MG A327496-10G
103825-468EA 37 USD
103825-468 103836-418 76839-384 76839-386 77002-980 77672-686
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride 97%
[1,4-Bis(diphenylphosphino)butane]palladium(II) dichloride

Precaución: for Research Use Only

Formula: C₂₈H₂₈Cl₂P₂Pd
MW: 603.80 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02093437
CAS Number: 29964-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR