1,4-Butanesultone 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Butane sultone

A161994-100G A161994-10G A161994-500G A161994-5G A161994-1000G A161994-25G
103808-904EA 180 USD
103808-904 103843-804 77191-118 77191-120 77191-122 77300-568 77319-826 77319-828
1,4-Butanesultone 98%
1,4-Butanesultone

Precaución: Inert atmosphere, room temperature

Formula: C₄H₈O₃S
MW: 136.17 g/mol
Boiling Pt: 299.9 °C (760 mm Hg)
Melting Pt: 12…15 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006584
CAS Number: 1633-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR