2-Methyl-1,4-phenylene bis{4-[3-(acryloyloxy)propoxy]benzoate} 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Bis-[4-(3-acryloyloxypropyloxy)benzoyloxy]-2-methylbenzene

A255324-1G A255324-25G A255324-100MG A255324-100G A255324-500G A255324-10G
103812-186EA 9 USD
103812-186 103852-042 77091-502 77091-504 77193-728 77550-622 77568-476 77593-598
2-Methyl-1,4-phenylene bis{4-[3-(acryloyloxy)propoxy]benzoate} 98%
2-Methyl-1,4-phenylene bis{4-[3-(acryloyloxy)propoxy]benzoate}

Precaución: For research use only.

Formula: C₃₃H₃₂O₁₀
MW: 588.601 g/mol
Boiling Pt: 733.5±60.0 °C (760 mm Hg)
Flash Pt: 303.9 °C
Storage Temperature: Ambient
MDL Number: MFCD09743681
CAS Number: 174063-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR