1,4-Bis[[(8α,9R)-10,11-dihydro-6′-methoxycinchonan-9-yl]oxy]-9,10-anthracenedione 96%

Supplier: Ambeed
Warning

Synonyms: (DHQ)2AQN

A302109-250MG A302109-100MG
103814-326EA 126 USD
103814-326 103835-482 77166-980 77683-898
1,4-Bis[[(8α,9R)-10,11-dihydro-6′-methoxycinchonan-9-yl]oxy]-9,10-anthracenedione 96%
1,4-Bis[[(8α,9R)-10,11-dihydro-6′-methoxycinchonan-9-yl]oxy]-9,10-anthracenedione

Precaución: For research use only.

Formula: C₅₄H₅₆N₄O₆
MW: 857.04 g/mol
Melting Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD29075644
CAS Number: 176097-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR