1,4-Bis(methoxymethyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: p-Xylene Glycol Dimethyl Ether

A100711-100G A100711-10G A100711-25G A100711-500G A100711-5G
103807-764EA 239 USD
103807-764 77188-528 77188-530 77188-532 77188-534 77745-766
1,4-Bis(methoxymethyl)benzene 98%
1,4-Bis(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Boiling Pt: 235 °C (760 mm Hg)
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00025903
CAS Number: 6770-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR