1,4-Benzenediboronic acid bis(pinacol)ester 97%

Supplier: Ambeed
Warning

Synonyms: 1,4-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,4-Phenylenediboronic acid, pinacol ester

A199169-100G A199169-10G A199169-250MG A199169-500G A199169-1G A199169-25G A199169-5G
103817-076EA 6 USD
103817-076 103847-700 76814-196 76814-198 76814-200 76814-202 77085-740 77085-742 77085-744
1,4-Benzenediboronic acid bis(pinacol)ester 97%
1,4-Benzenediboronic acid bis(pinacol)ester

Precaución: for Research Use Only

Formula: C₁₈H₂₈B₂O₄
Boiling Pt: 416.3 °C (760 mm Hg)
Melting Pt: 241…245 °C
Flash Pt: 205.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08276852
CAS Number: 99770-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR