1,4-Bis(2-chloroethyl)piperazine 97%

Supplier: Ambeed
Warning

Synonyms: PIPERAZINE, 1,4-BIS(2-CHLOROETHYL)-

A236067-1G A236067-5G A236067-250MG
103811-068EA 42 USD
103811-068 103850-838 77089-876 77089-878 77683-699
1,4-Bis(2-chloroethyl)piperazine 97%
1,4-Bis(2-chloroethyl)piperazine

Precaución: For research use only.

Formula: C₈H₁₆Cl₂N₂
MW: 211.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01703917
CAS Number: 1009-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR