1,4-Benzodioxane-6-boronic acid 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,3-Dihydro-1,4-benzodioxine-6-boronic acid
A656685-25G
A656685-5G
A656685-100G
A656685-10G
A656685-250MG
103840-422EA
21
USD
103840-422
76844-610
76844-612
77254-740
77254-742
77254-744
1,4-Benzodioxane-6-boronic acid 96%
1,4-Benzodioxane-6-boronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BO₄ MW: 179.97 g/mol Boiling Pt: 361 °C (760 mm Hg) Melting Pt: 187 °C Flash Pt: 172.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009696 CAS Number: 164014-95-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...