1,4,8,11-Tetraazacyclotetradecane 97%

Supplier: Ambeed
Warning

A112684-10G A112684-250MG A112684-25G A112684-1G A112684-5G
103798-074EA 12 USD
103798-074 103823-912 76796-934 76796-936 76796-938 77310-724 77310-726
1,4,8,11-Tetraazacyclotetradecane 97%
1,4,8,11-Tetraazacyclotetradecane

Precaución: for Research Use Only

Formula: C₁₀H₂₄N₄
MW: 200.33 g/mol
Boiling Pt: 321.1 °C (760 mm Hg)
Melting Pt: 184…186 °C
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00005105
CAS Number: 295-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR