1,4,7-Trimethyl-1,4,7-triazacyclononane 97% stabilized
Supplier: Ambeed
|
Danger
|
Synonyms:
1,4,7-Trimethyl-1,4,7-triazonane
Stabilization:
Stabilised with sodium hydrogen carbonate (NaHCO₃)
A418725-1G
A418725-25G
A418725-5G
A418725-100G
A418725-10G
103827-360EA
9
USD
103827-360
103838-404
76842-012
76842-014
76842-016
77196-186
77196-188
1,4,7-Trimethyl-1,4,7-triazacyclononane 97% stabilized
1,4,7-Trimethyl-1,4,7-triazacyclononane
Precaución: for Research Use Only
Formula:
C₉H₂₁N₃ MW: 171.28 g/mol Boiling Pt: 207.8 °C (760 mm Hg) Flash Pt: 67 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00012359 CAS Number: 96556-05-7 UN: 3267 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...