1,4,7,10-Tetraazacyclododecane 98%

Supplier: Ambeed
Danger

Synonyms: Cyclen

A109765-100G A109765-10G A109765-25G A109765-500G A109765-1G A109765-5G
103797-558EA 8 USD
103797-558 103823-364 77093-928 77093-930 77093-932 77093-934 77283-882 77310-176
1,4,7,10-Tetraazacyclododecane 98%
1,4,7,10-Tetraazacyclododecane

Precaución: for Research Use Only

Formula: C₈H₂₀N₄
MW: 172.27 g/mol
Boiling Pt: 283.8 °C (760 mm Hg)
Melting Pt: 108…113 °C
Flash Pt: 129.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00066281
CAS Number: 294-90-6
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR