1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H,4H,5H,6H-Cyclopenta[c]pyrazole-3-carboxylic acid
A607385-10G
A607385-250MG
A607385-1G
A607385-5G
103840-150EA
129
USD
103840-150
77170-174
77170-176
77307-936
77307-938
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid 98%
1,4,5,6-Tetrahydrocyclopenta[c]pyrazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Boiling Pt: 430.7 °C (760 mm Hg) Melting Pt: 291 °C at N/A mmHg Flash Pt: 214.3 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01248822 CAS Number: 5932-32-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...