1,3-Phenyldiboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Benzenediboronic acid, m-Phenylenebisboronic acid, 1,3-Phenylenediboronic acid, 1,3-Phenyldiboric acid

A135430-100G A135430-250MG A135430-1G A135430-25G A135430-5G A135430-10G
103802-152EA 17 USD
103802-152 77095-190 77095-192 77314-970 77314-972 77314-974 77566-654
1,3-Phenyldiboronic acid 97%
1,3-Phenyldiboronic acid

Precaución: For research use only.

Formula: C₆H₈B₂O₄
MW: 165.75 g/mol
Boiling Pt: 456.4 °C (760 mm Hg)
Melting Pt: >300 °C
Flash Pt: 229.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03092915
CAS Number: 4612-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR