1,3-Di-tert-butyltoluene 98%
Supplier: Ambeed
|
Warning
|
A223786-1G
A223786-25G
A223786-100G
103819-062EA
5
USD
103819-062
103849-944
77088-566
77088-568
77275-554
1,3-Di-tert-butyltoluene 98%
1,3-Di-tert-butyltoluene
Precaución: for Research Use Only
Formula:
C₁₅H₂₄ Boiling Pt: 244 °C (760 mm Hg) Melting Pt: 31…32 °C Flash Pt: 91.11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00026300 CAS Number: 15181-11-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...