1,3-Dimethyl-1H-pyrazole-5-boronic acid pinacol ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
A131198-100MG
A131198-250MG
A131198-1G
A131198-5G
103801-540EA
21
USD
103801-540
76972-066
76972-068
77314-010
77314-012
1,3-Dimethyl-1H-pyrazole-5-boronic acid pinacol ester 97%
1,3-Dimethyl-1H-pyrazole-5-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₁H₁₉BN₂O₂ MW: 222.0918 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD05664574 CAS Number: 847818-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...