1,3-Dimethyl-1H-pyrazol-5-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dimethyl-1H-pyrazole-5-carboxaldehyde, 1,3-Dimethyl-1H-pyrazole-5-carbaldehyde

A470610-100MG A470610-10G A470610-1G A470610-5G A470610-250MG
103828-024EA 37 USD
103828-024 103839-098 76774-274 76774-276 76843-008 76843-010 77626-940
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde 98%
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.1400 g/mol
Boiling Pt: 227.7 °C (760 mm Hg)
Melting Pt: 40…42 °C
Storage Temperature: Ambient
MDL Number: MFCD02681907
CAS Number: 25016-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR