1,3-Dimethyl-1H-pyrazol-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dimethyl-1H-pyrazole-4-carbaldehyde

A123145-10G A123145-25G A123145-1G A123145-5G A123145-250MG
103800-350EA 27 USD
103800-350 77026-584 77026-586 77297-928 77297-930 77599-796
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde 97%
1,3-Dimethyl-1H-pyrazol-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MW: 124.1405 g/mol
Flash Pt: 92.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159635
CAS Number: 25016-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR