1,3-Dimethyl-1H-pyrazole 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Dimethylpyrazole

A105054-100G A105054-10G A105054-500G A105054-1G A105054-5G A105054-25G
103796-828EA 9 USD
103796-828 103822-554 77188-726 77188-728 77188-730 77309-424 77309-426 77592-118
1,3-Dimethyl-1H-pyrazole 98%
1,3-Dimethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₈N₂
MW: 96.1304 g/mol
Boiling Pt: 153 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00036665
CAS Number: 694-48-4
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR