1,3-Dimethoxypropan-2-one 96%

Supplier: Ambeed
Danger

A174014-5G A174014-10G A174014-250MG
103806-900EA 45 USD
103806-900 103845-200 77082-486 77191-506 77191-508
1,3-Dimethoxypropan-2-one 96%
1,3-Dimethoxypropan-2-one

Precaución: for Research Use Only

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 162.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00270306
CAS Number: 18664-32-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR