1,3-Dimethoxypropan-2-ol 98%

Supplier: Ambeed
Warning

A239259-1G A239259-5G A239259-100G A239259-10G A239259-500G A239259-25G
103811-270EA 6 USD
103811-270 103851-068 77090-206 77090-208 77193-280 77193-282 77193-284 77304-146
1,3-Dimethoxypropan-2-ol 98%
1,3-Dimethoxypropan-2-ol

Precaución: for Research Use Only

Formula: C₅H₁₂O₃
MW: 120.1500 g/mol
Boiling Pt: 169 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD01076303
CAS Number: 623-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR