1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,3-Bis(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium tetrafluoroborate, 1,3-Bis(2,4,6-trimethylphenyl)imidazolinium tetrafluoroborate, SIMes-HBF4
A234669-5G
A234669-250MG
103810-968EA
24
USD
103810-968
103850-724
77089-718
77193-172
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%
1,3-Dimesityl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₁H₂₇BF₄N₂ Melting Pt: 290…296 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684542 CAS Number: 245679-18-9 UN: 1759 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...