1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride
A224297-1G
A224297-5G
A224297-100G
A224297-10G
A224297-250MG
103819-096EA
6
USD
103819-096
103849-986
77088-630
77088-632
77192-972
77192-974
77192-976
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride 98%
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
Precaución: for Research Use Only
Formula:
C₂₁H₂₅ClN₂ MW: 340.89 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD02684541 CAS Number: 141556-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...