1,3-Dihydroxyacetone 98%

Supplier: Ambeed
Warning

Synonyms: Dihydroxyacetone

A159350-100G A159350-10G A159350-500G A159350-25G
103808-870EA 83 USD
103808-870 76798-934 76798-936 76798-938 77319-342
1,3-Dihydroxyacetone 98%
1,3-Dihydroxyacetone

Precaución: for Research Use Only

Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 213.7 °C (760 mm Hg)
Melting Pt: 75…80 °C
Flash Pt: 97.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004670
CAS Number: 96-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR