5-Amino-1,3-dihydrobenzo[c]furan 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dihydroisobenzofuran-5-amine, 5-Amino-1,3-dihydro-2-benzofuran, 1,3-Dihydro-2-benzofuran-5-amine

A159393-250MG A159393-1G A159393-5G
103805-250EA 55 USD
103805-250 103843-468 77191-020 77319-350 77319-352
5-Amino-1,3-dihydrobenzo[c]furan 97%
5-Amino-1,3-dihydrobenzo[c]furan

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₈H₉NO
MW: 135.163 g/mol
Boiling Pt: 295.9 °C (760 mm Hg)
Flash Pt: 156.803 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05864561
CAS Number: 61964-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR