3,5-Difluorotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Difluoro-5-methylbenzene

A635269-1G A635269-25G A635269-5G A635269-10G
103840-306EA 22 USD
103840-306 76844-474 76844-476 76844-478 77117-542
3,5-Difluorotoluene 98%
3,5-Difluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 117.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00797323
CAS Number: 117358-51-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR