1,3-Difluoro-5-methyl-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A113113-100MG A113113-250MG
103798-164EA 131 USD
103798-164 103824-004 76970-990 76970-992
1,3-Difluoro-5-methyl-2-nitrobenzene 98%
1,3-Difluoro-5-methyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD15527695
CAS Number: 932373-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR