1,3-Difluoro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoronitrobenzene

A180962-25G A180962-5G A180962-100G A180962-10G A180962-500G A180962-1G
103807-574EA 9 USD
103807-574 103845-936 77083-448 77083-450 77287-202 77287-204 77287-206 77301-700
1,3-Difluoro-2-nitrobenzene 98%
1,3-Difluoro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 220.1 °C (760 mm Hg)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00192035
CAS Number: 19064-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR