1,3-Difluoro-2-methyl-5-nitrobenzene 97%

Supplier: Ambeed
Warning

A132644-250MG A132644-1G A132644-5G
103801-754EA 132 USD
103801-754 76983-806 77298-566 77298-568
1,3-Difluoro-2-methyl-5-nitrobenzene 97%
1,3-Difluoro-2-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.11 g/mol
Boiling Pt: 231 °C (760 mm Hg)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD03094192
CAS Number: 170572-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR