1,3-difluoro-2-methyl-4-nitrobenzene 97%

Supplier: Ambeed
Danger

Synonyms: 2,6-Difluoro-3-nitrotoluene, 1,3-Difluoro-2-methyl-4-nitrobenzene

A808733-10G A808733-250MG A808733-1G A808733-25G A808733-5G
103841-082EA 46 USD
103841-082 76791-662 76791-664 76845-330 76845-332 76845-334
1,3-difluoro-2-methyl-4-nitrobenzene 97%
1,3-difluoro-2-methyl-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.119 g/mol
Boiling Pt: 237.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11519030
CAS Number: 79562-49-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR