1,3-Dicyclohexylimidazolium chloride 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Dicyclohexyl-1H-imidazol-3-ium chloride

A140177-10G A140177-250MG A140177-1G A140177-25G A140177-5G
103802-842EA 11 USD
103802-842 77190-368 77190-370 77315-958 77315-960 77315-962
1,3-Dicyclohexylimidazolium chloride 97%
1,3-Dicyclohexylimidazolium chloride

Precaución: for Research Use Only

Formula: C₁₅H₂₅ClN₂
MW: 268.8254 g/mol
Melting Pt: 101…144 °C
Storage Temperature: Ambient
MDL Number: MFCD04973309
CAS Number: 181422-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR