1,3-Dichloroisoquinoline 98%

Supplier: Ambeed
Warning

A194398-1G A194398-5G A194398-10G A194398-250MG A194398-25G
103816-652EA 11 USD
103816-652 103847-262 77085-178 77085-180 77111-914 77111-916 77302-422
1,3-Dichloroisoquinoline 98%
1,3-Dichloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅Cl₂N
Melting Pt: 121…122 °C
Flash Pt: 182.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00034750
CAS Number: 7742-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR