α,3,5-Trichlorotoluene 96%

Supplier: Ambeed
Danger

Synonyms: 1-(Chloromethyl)-3,5-dichlorobenzene, 1,3-Dichloro-5-(chloromethyl)benzene, 3,5-Dichlorobenzyl chloride

A174518-1G A174518-25G A174518-5G A174518-10G A174518-250MG
103806-952EA 22 USD
103806-952 103845-252 77082-544 77082-546 77082-548 77096-582 77096-584
α,3,5-Trichlorotoluene 96%
α,3,5-Trichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Cl₃
MW: 195.47
Boiling Pt: 248.3 °C (760 mm Hg)
Melting Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060278
CAS Number: 3290-06-0
UN: 3261
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR