3,5-Dibromoanisole 98%

Supplier: Ambeed
Warning

A596910-100G A596910-10G A596910-500G A596910-1G A596910-5G A596910-25G
103828-956EA 5 USD
103828-956 103840-094 76775-160 76775-162 76775-164 76844-244 76844-246 77307-910
3,5-Dibromoanisole 98%
3,5-Dibromoanisole

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
Boiling Pt: 124 °C (760 mm Hg)
Melting Pt: 34…38 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02258848
CAS Number: 74137-36-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR