1,3-Dibromo-5-iodobenzene 97%

Supplier: Ambeed
Warning

A220356-1G A220356-5G A220356-10G A220356-25G A220356-100G
103818-850EA 8 USD
103818-850 103849-732 77088-318 77088-320 77192-872 77303-550 77746-309
1,3-Dibromo-5-iodobenzene 97%
1,3-Dibromo-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂I
Boiling Pt: 302.6 °C (760 mm Hg)
Melting Pt: 124.4…124.7 °C
Flash Pt: 136.856 °C
Storage Temperature: Ambient
MDL Number: MFCD07778996
CAS Number: 19752-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR