3,5-Dibromo-α,α,α-trifluorotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromobenzotrifluoride, 1,3-Dibromo-5-(trifluoromethyl)benzene

A372255-1G A372255-25G A372255-5G A372255-100G A372255-10G A372255-250MG
103826-500EA 9 USD
103826-500 103837-518 76840-770 76840-772 76840-774 77278-432 77278-434 77278-436
3,5-Dibromo-α,α,α-trifluorotoluene 98%
3,5-Dibromo-α,α,α-trifluorotoluene

Precaución: for Research Use Only

Formula: C₇H₃Br₂F₃
MW: 303.90 g/mol
Boiling Pt: 200.1 °C (760 mm Hg)
Flash Pt: 74.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00526219
CAS Number: 401-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR