1,3-Dibromo-5-(trifluoromethoxy)benzene 95%

Supplier: Ambeed
Warning

A136720-100G A136720-500G A136720-25G
77190-220EA 85.49 USD
77190-220 77190-222 77315-262
1,3-Dibromo-5-(trifluoromethoxy)benzene 95%
1,3-Dibromo-5-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃Br₂F₃O
MW: 319.90 g/mol
Boiling Pt: 211.7 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00075280
CAS Number: 207226-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR