α,3,5-Tribromotoluene 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
3,5-Dibromobenzyl bromide, 1,3-Dibromo-5-(bromomethyl)benzene, alpha,3,5-Tribromotoluene
A877740-25G
A877740-5G
A877740-10G
A877740-1G
103841-306EA
11
USD
103841-306
76845-572
76845-574
77118-690
77308-484
α,3,5-Tribromotoluene 98%
α,3,5-Tribromotoluene
Precaución: for Research Use Only
Formula:
C₇H₅Br₃ MW: 328.83 Boiling Pt: 173 Melting Pt: 93…97 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00052415 CAS Number: 56908-88-4 UN: 2921 ADR: 8,4.1 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...